missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(4-Formylaminobutyryl)pyridine, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | 3-(4-Formylaminobutyryl)pyridine |
| CAS | 887355-56-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C10H12N2O2 |
| Quantity | 100 mg |
| Synonym | Nicotine Impurity 1 |
| InChI Formula | InChI=1 S/C10H12N2O2/c13-8-12-6-2-4-10(14)9-3-1-5-11-7-9/h1,3,5,7-8 H,2,4,6H2,(H,12,13) |
| SMILES | O=CNCCCC(=O)c1cccnc1 |
| Molecular Weight (g/mol) | 192.21 |
| Formula Weight | 192.0899 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction