Learn More
3-(4-Fluorophenyl)-3-oxopropanenitrile, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 4640-67-9 |
| Molecular Formula | C9H6FNO |
| MDL Number | MFCD00662062 |
| Quantity | 10 g |
| SMILES | N#CCC(C1=CC=C(F)C=C1)=O |
| Molecular Weight (g/mol) | 163.15 |
| Percent Purity | 98% |
| Chemical Name or Material | 3-(4-Fluorophenyl)-3-oxopropanenitrile |