Learn More
3-(4-Fluorobenzoyl)-propionic acid, Lipomed
Product Code. 30461883
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30461883 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30461883 Supplier Lipomed Supplier No. LPMFPA1879FA50
 Restricted Item
Estimated Shipment: 30-11-2026
Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.
3-(4-Fluorobenzoyl)-propionic acid
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-(4-Fluorophenyl)-4-oxobutanoic Acid
CAS 366-77-8
Analyte or Component Names 4-(4-Fluorophenyl)-4-oxobutanoic Acid
Shipping Condition Room Temperature
Molecular Formula C10 H9 F O3
Synonym Benzenebutanoic acid, 4-fluoro-γ-oxo- (9 CI, ACI), 4-Fluoro-γ-oxobenzenebutanoic acid (ACI), Propionic acid, 3-(p-fluorobenzoyl)- (7 CI, 8 CI), 3-(4-Fluorobenzoyl)propanoic acid, 3-(4-Fluorobenzoyl)propionic acid, 3-(p-Fluorobenzoyl)propionic acid, 4-(4-Fluorophenyl)-4-oxobutanoic acid, 4-(4-Fluorophenyl)-4-oxobutyric acid, 4-Fluorobenzoylpropionic acid, 4-Oxo-4-(p-fluorophenyl)butyric acid, Ba 2833, NSC 408180, β-(4-Fluorobenzoyl)propionic acid, β-(p-Fluorobenzoyl)propionic acid, γ-(4-Fluorophenyl)-γ-oxobutanoic acid
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES OC(=O)CCC(=O)c1ccc(F)cc1
IUPAC Name 4-(4-fluorophenyl)-4-oxobutanoic acid
Industry Type Pharmaceutical
Molecular Weight (g/mol) 196.175
Quantity 50 mg
Formula Weight 196.054 g/mol