Learn More
3,4-Dimethylfuran, TRC
Specifications
Specifications
| CAS | 20843-07-6 |
| Molecular Formula | C6 H8 O |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C6H8O/c1-5-3-7-4-6(5)2/h3-4H,1-2H3 |
| SMILES | Cc1cocc1C |
| IUPAC Name | 3,4-dimethylfuran |
| Molecular Weight (g/mol) | 96.13 |
| Formula Weight | 96.0575 |
| Chemical Name or Material | 3,4-Dimethylfuran |