missing translation for 'onlineSavingsMsg'
Learn More

3',4'-Dimethoxyflavone, 97%, Thermo Scientific™

Product Code. 11369015
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 11369015

Brand: Thermo Scientific Alfa Aesar L14160.MC

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4143-62-8
Molecular Formula C17H14O4
Molecular Weight (g/mol) 282.295
MDL Number MFCD00143009
InChI Key ZGHORMOOTZTQFL-UHFFFAOYSA-N
Synonym 3',4'-dimethoxyflavone, 2-3,4-dimethoxyphenyl-4h-chromen-4-one, 2-3,4-dimethoxyphenyl chromen-4-one, chembl91153, 4h-1-benzopyran-4-one, 2-3,4-dimethoxyphenyl, 3',4'-dmf, pubchem9847, spectrum_001039, acmc-1apwv, specplus_000555
PubChem CID 688674
IUPAC Name 2-(3,4-dimethoxyphenyl)chromen-4-one
SMILES COC1=C(C=C(C=C1)C2=CC(=O)C3=CC=CC=C3O2)OC

Specifications

Melting Point 154°C to 155°C
Quantity 100 mg
Beilstein 236749
Formula Weight 282.3
Percent Purity 97%
Chemical Name or Material 3',4'-Dimethoxyflavone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.