Learn More
3',4'-Dimethoxyflavone, 97%, Thermo Scientific™
Product Code. 11359015
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Product Code. Quantity unitSize
11359015 500 mg 500mg
11369015 100 mg 100mg
2 options
Product Code. 11359015
Brand: Thermo Scientific Alfa Aesar L14160.ME
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4143-62-8
Molecular Formula C17H14O4
Molecular Weight (g/mol) 282.295
MDL Number MFCD00143009
InChI Key ZGHORMOOTZTQFL-UHFFFAOYSA-N
Synonym 3',4'-dimethoxyflavone, 2-3,4-dimethoxyphenyl-4h-chromen-4-one, 2-3,4-dimethoxyphenyl chromen-4-one, chembl91153, 4h-1-benzopyran-4-one, 2-3,4-dimethoxyphenyl, 3',4'-dmf, pubchem9847, spectrum_001039, acmc-1apwv, specplus_000555
PubChem CID 688674
IUPAC Name 2-(3,4-dimethoxyphenyl)chromen-4-one
SMILES COC1=C(C=C(C=C1)C2=CC(=O)C3=CC=CC=C3O2)OC
Specifications
Melting Point 154°C to 155°C
Quantity 500 mg
Beilstein 236749
Formula Weight 282.3
Percent Purity 97%
Chemical Name or Material 3',4'-Dimethoxyflavone