missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxybenzyl alcohol, 97%

Product Code. p-7021168
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11435403 25 g 25g
11445403 100 g 100g
11455403 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11435403 Supplier Thermo Scientific Alfa Aesar Supplier No. A13396.14
Add to basket
This item is not returnable. View return policy

CAS: 93-03-8 | C9H12O3 | 168.192 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
3,4-Dimethoxybenzyl alcohol used as the fuel of the microbial fuel cell (MFC) to generate power. Veratrole alcohol is the raw material for the synthesis of cyclotriveratrylene which is used in host-guest chemistry.

Solubility
Fully miscible with water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-03-8
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.192
MDL Number MFCD00004638
InChI Key OEGPRYNGFWGMMV-UHFFFAOYSA-N
Synonym 3,4-dimethoxybenzyl alcohol, veratryl alcohol, 3,4-dimethoxyphenyl methanol, benzenemethanol, 3,4-dimethoxy, 3,4-dimethoxyphenylmethyl alcohol, veratrole alcohol, 3,4-dimethoxybenzenemethanol, unii-mb4t4a711h, 3,4-dimethoxyphenyl-methanol
PubChem CID 7118
ChEBI CHEBI:62150
IUPAC Name (3,4-dimethoxyphenyl)methanol
SMILES COC1=C(C=C(C=C1)CO)OC

Specifications

Melting Point ∼22°C
Density 1.176
Boiling Point 297°C to 298°C
Flash Point >110°C (230°F)
Refractive Index 1.553
Quantity 25 g
Beilstein 639388
Solubility Information Fully miscible with water.
Formula Weight 168.19
Percent Purity 97%
Chemical Name or Material 3,4-Dimethoxybenzyl alcohol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.