Learn More
3,4-Dimethoxy-3-cyclobutene-1,2-dione, 98%
Product Code. p-7024032
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 11473707
Brand: Thermo Scientific Alfa Aesar A18186.03
Request Bulk or Custom Format
CAS: 5222-73-1 | C6H6O4 | 142.11 g/mol
3,4-Dimethoxy-3-cyclobutene-1,2-dione (Dimethyl squarate) finds it uses in the synthesis of chiral squaramides, highly enantioselective catalyst for the Friedel-Crafts reactions of indoles, 3-(hydroxyamino)-4-methoxy-3-cyclobutene-1,2-dione, squarate derivatives of the O-SP-core antigens, methyl squarate derivative of the Ogawa O-SP-core antigen, o-quinodimethanes, benzocyclobutenes, quinones. It is employed as a starting material in the preparation of o-quinodimethanes, benzocyclobutenes, and quinones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,4-Dimethoxy-3-cyclobutene-1,2-dione (Dimethyl squarate) finds it uses in the synthesis of chiral squaramides, highly enantioselective catalyst for the Friedel-Crafts reactions of indoles, 3-(hydroxyamino)-4-methoxy-3-cyclobutene-1,2-dione, squarate derivatives of the O-SP-core antigens, methyl squarate derivative of the Ogawa O-SP-core antigen, o-quinodimethanes, benzocyclobutenes, quinones. It is employed as a starting material in the preparation of o-quinodimethanes, benzocyclobutenes, and quinones.
Solubility
Very soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5222-73-1
Molecular Formula C6H6O4
Molecular Weight (g/mol) 142.11
MDL Number MFCD00101316
InChI Key SZBNZTGCAMLMJY-UHFFFAOYSA-N
PubChem CID 123227
IUPAC Name 3,4-dimethoxycyclobut-3-ene-1,2-dione
SMILES COC1=C(C(=O)C1=O)OC
Specifications
Melting Point 53°C to 56°C
Quantity 1 g
Synonym 3,4-dimethoxy-3-cyclobutene-1,2-dione, dimethyl squarate, squaric acid dimethyl ester, 3-cyclobutene-1,2-dione, 3,4-dimethoxy, 3,4-dimethoxy-cyclobut-3-ene-1,2-dione, dimethoxycyclobut-3-ene-1,2-dione, 3-cyclobutene-1,2-dione,3,4-dimethoxy, 3,4-dimethoxycyclobut-3-en-1,2-dion, dimethoxycyclobutenedione, pubchem13709
Solubility Information Very soluble in water.
Formula Weight 142.1
Percent Purity 98%
Chemical Name or Material 3,4-Dimethoxy-3-cyclobutene-1,2-dione