Learn More
3',4'-Dihydroxyphenylacetone, TRC
Product Code. 30007289
Change view
Click to view available options
Quantity:
25 mg
100 mg
500 mg
Unit Size:
100mg
25mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007289 100 mg 100mg
30003518 500 mg 500mg
30008703 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30007289 Supplier Toronto Research Chemicals Supplier No. TRCD454300100MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2503-44-8
Molecular Formula C9 H10 O3
Molecular Weight (g/mol) 166.17
Synonym 1-(3,4-Dihydroxyphenyl)-2-propanone, 2-Propanone, (3,4-dihydroxyphenyl)- (6CI,7CI), 1-(3,4-Dihydroxyphenyl)-2-propanone, 1-(3',4'-Dihydroxyphenyl)-2-propanone, 3,4-Dihydroxyphenylacetone, Carbidopa Imp. G (EP)
IUPAC Name 1-(3,4-dihydroxyphenyl)propan-2-one
SMILES CC(=O)Cc1ccc(O)c(O)c1
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C9H10O3/c1-6(10)4-7-2-3-8(11)9(12)5-7/h2-3,5,11-12H,4H2,1H3
Formula Weight 166.063
Percent Purity >95%
Chemical Name or Material 3,4-Dihydroxyphenylacetone
Grade HPLC