Learn More
3,4'-Dihydroxyflavone, MedChemExpress
Product Code. 16475386
Encompass_Preferred
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. 16475386
Brand: Medchem Express HY111802
MedChemExpress 3,4'-Dihydroxyflavone (3,4'-DHF) is an oral active flavonoid with antiviral activity against Influenza A virus.
3,4'-Dihydroxyflavone (3,4'-DHF) is an oral active flavonoid with antiviral activity against Influenza A virus.
Specifications
Chemical Name or Material 3,4'-Dihydroxyflavone
Physical Form Solid
CAS 14919-49-4
Color Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C15H10O4
For Use With (Application) COVID-19-anti-virus
Synonym 3,4'-DHF
Solubility Information DMSO : ≥ 155 mg/mL (609.66 mM)
SMILES O=C1C(O)=C(C2=CC=C(O)C=C2)OC3=C1C=CC=C3
Molecular Weight (g/mol) 254.24
Formula Weight 254.24
Percent Purity 98.15%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.