missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorothiobenzamide, 97%, Thermo Scientific™

Catalog Number 15460828
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
317319-14-5
C7H5F2NS
173.18
MFCD04036083
BVZJSQFOGOEVMF-UHFFFAOYSA-N
16771464
3,4-difluorobenzenecarbothioamide
NC(=S)C1=CC(F)=C(F)C=C1
This item is not returnable. View return policy
317319-14-5
C7H5F2NS
173.18
MFCD04036083
BVZJSQFOGOEVMF-UHFFFAOYSA-N
16771464
3,4-difluorobenzenecarbothioamide
NC(=S)C1=CC(F)=C(F)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Synonym 3,4-difluorobenzene-1-carbothioamide, 3,4-difluorothiobenzamide, 3,4-difluorobenzothioamide, 3,4-difluoro-thiobenzamide, benzenecarbothioamide,3,4-difluoro, benzenecarbothioamide, 3,4-difluoro, benzenecarbothioamide, 3,4-difluoro-9ci
Formula Weight 173.18
Percent Purity 97%
Chemical Name or Material 3,4-Difluorothiobenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'