Learn More
(3,4-Dichlorophenyl)methanesulfonyl chloride, 90%, Thermo Scientific™
Product Code. 10064582
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10064582 250 mg 250mg
10514373 1 g 1g
10198824 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10064582 Supplier Thermo Scientific Maybridge Supplier No. MO00820CB
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Chemical Identifiers
CAS 85952-30-3
Molecular Formula C7H5Cl3O2S
Molecular Weight (g/mol) 259.525
MDL Number MFCD01631929
InChI Key DRYGYFAFCNDLEJ-UHFFFAOYSA-N
Synonym 3,4-dichlorophenyl methanesulfonyl chloride, 3,4-dichloro-phenyl-methanesulfonyl chloride, 3,4-dichlorophenyl methanesulphonyl chloride, 3,4-dichlorophenyl methyl sulfonyl chloride, 3,4-dichlorobenzylsulfonyl chloride, 3,4-dichlorophenyl methyl chlorosulfone, 3,4-dichloro-alpha-toluenesulfonyl chloride, 3,4-dichlorophenyl-methanesulfonyl chloride, benzenemethanesulfonyl chloride, 3,4-dichloro, 3,4-dichlorophenyl methyl sulphonyl chloride
PubChem CID 2794622
IUPAC Name (3,4-dichlorophenyl)methanesulfonyl chloride
SMILES C1=CC(=C(C=C1CS(=O)(=O)Cl)Cl)Cl
Specifications
CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 259.54
Percent Purity 90%
Chemical Name or Material (3, 4-Dichlorophenyl)methanesulfonyl chloride