Learn More
(3,4-Dichlorophenyl)methanesulfonyl chloride, 90%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
CAS Min % | 90.0 |
Assay Percent Range | 90% |
Packaging | Amber glass bottle |
Quantity | 250 mg |
Formula Weight | 259.54 |
Percent Purity | 90% |
Chemical Name or Material | (3, 4-Dichlorophenyl)methanesulfonyl chloride |