Learn More
3-(4-Chlorophenoxy)-1,2-propanediol, 99%
Product Code. 11403637
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
Product Code. Quantity unitSize
11403637 100 g 100g
11493627 25 g 25g
11413637 500 g 500g
3 options
Product Code. 11403637
Brand: Thermo Scientific Alfa Aesar A17386.22
Request Bulk or Custom Format
CAS: 104-29-0 | C9H11ClO3 | 202.634 g/mol
It inhibits IgE-mediated histamine release. 3-(4-Chlorophenoxy)-1,2-propanediol is also used as an antimycotic agent. It is a centrally acting muscle relaxant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It inhibits IgE-mediated histamine release. 3-(4-Chlorophenoxy)-1,2-propanediol is also used as an antimycotic agent. It is a centrally acting muscle relaxant.
Solubility
Slightly soluble in water.
Notes
Store at 78-82°C. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 104-29-0
Molecular Formula C9H11ClO3
Molecular Weight (g/mol) 202.634
MDL Number MFCD00021990
InChI Key MXOAEAUPQDYUQM-UHFFFAOYSA-N
Synonym chlorphenesin, 3-4-chlorophenoxy-1,2-propanediol, 3-4-chlorophenoxy propane-1,2-diol, chlorophenesin, chlorphenesinum, adermykon, gecophen, demykon, mycil, chlorphenesine
PubChem CID 7697
ChEBI CHEBI:3642
IUPAC Name 3-(4-chlorophenoxy)propane-1,2-diol
SMILES C1=CC(=CC=C1OCC(CO)O)Cl
Specifications
Melting Point 78°C to 82°C
Quantity 100 g
Solubility Information Slightly soluble in water.
Formula Weight 202.64
Percent Purity 99%
Chemical Name or Material 3-(4-Chlorophenoxy)-1,2-propanediol
compliance-icons
Product Identifier
  • 3-(4-Chlorophenoxy)-1,2-propanediol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Chlorphenesin