missing translation for 'onlineSavingsMsg'
Learn More

3,4,9,10-Perylenetetracarboxylic 3,4:9,10-Dianhydride, Sigma-Aldrich™

Product Code. 15821301
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15821301 100 g 100g
15811301 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15821301 Supplier Sigma Aldrich Supplier No. 8209700100
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128-69-8
Grade Research
Molecular Weight (g/mol) 392.32

Specifications

Melting Point >350°C
MDL Number MFCD00006916
Quantity 100 g
Synonym 3,4,9,10-Perylenetetracarboxylic 3,4:9,10-dianhydride, 3,4,9,10-Perylenetetracarboxylic 3,4:9,10-dianhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.