missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trihydroxybenzoic acid, 98%

Product Code. 11441347
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
This item is not returnable. View return policy

Product Code. 11441347

Brand: Thermo Scientific Alfa Aesar B24887.0B

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 149-91-7 | C7H6O5 | 170.12 g/mol

Organic acid is used as a standard for determining phenols in the Folin-Ciocalteau assay3,4,5-Trihydroxybenzoic acid is used as a cyclooxygease inhibitor substance, which is found in plants. In analytical chemistry, it is used as a standard to determine phenol content in various analytes. It acts as a starting material for the preparation of psychedelic alkaloid mescaline. Further, it is used to produce polyesters based on phloretic acid and gallic acid. In addition to this, it is used in the preparation of its esters such as methyl gallate, ethyl gallate, propyl gallate, octyl gallate and doceyl gallate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Organic acid is used as a standard for determining phenols in the Folin-Ciocalteau assay3,4,5-Trihydroxybenzoic acid is used as a cyclooxygease inhibitor substance, which is found in plants. In analytical chemistry, it is used as a standard to determine phenol content in various analytes. It acts as a starting material for the preparation of psychedelic alkaloid mescaline. Further, it is used to produce polyesters based on phloretic acid and gallic acid. In addition to this, it is used in the preparation of its esters such as methyl gallate, ethyl gallate, propyl gallate, octyl gallate and doceyl gallate.

Solubility
Soluble in water, alcohol, ether, glycerol and acetone. Insoluble in benzene, chloroform and petroleum ether.

Notes
Hygroscopic. Light sensitive. Incompatible with strong oxidizing agents, strong bases, acid anhydrides, acid chlorides, metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149-91-7
Molecular Formula C7H6O5
Molecular Weight (g/mol) 170.12
MDL Number MFCD00002510
InChI Key LNTHITQWFMADLM-UHFFFAOYSA-N
Synonym gallic acid, gallate, benzoic acid, 3,4,5-trihydroxy, gallic acid, tech., galop, pyrogallol-5-carboxylic acid, kyselina gallova, 3,4,5-trihydroxybenzoate, ccris 5523, kyselina gallova czech
PubChem CID 370
ChEBI CHEBI:30778
IUPAC Name 3,4,5-trihydroxybenzoic acid
SMILES OC(=O)C1=CC(O)=C(O)C(O)=C1

Specifications

Melting Point ∼251°C (decomposition)
Density 1.694
Quantity 1000 g
Beilstein 2050274
Sensitivity Hygroscopic; Light sensitive
Merck Index 144345
Solubility Information Soluble in water,alcohol,ether,glycerol and acetone. Insoluble in benzene,chloroform and petroleum ether.
Formula Weight 170.12
Percent Purity 0.99
Chemical Name or Material 3,4,5-Trihydroxybenzoic acid, anhydrous

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.