Learn More
3,4,5-Trichlorophenol, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 3,4,5-Trichlorophenol |
| CAS | 609-19-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C6 H3 Cl3 O |
| Quantity | 1 g |
| Synonym | Phenol','3,4,5-trichloro-, 3,4,5-Trichlorophenol, NSC 243667 |
| InChI Formula | InChI=1S/C6H3Cl3O/c7-4-1-3(10)2-5(8)6(4)9/h1-2,10H |
| SMILES | Oc1cc(Cl)c(Cl)c(Cl)c1 |
| IUPAC Name | 3,4,5-trichlorophenol |
| Molecular Weight (g/mol) | 197.45 |
| Show More |