Learn More
3-(3-Pyridyl)benzeneboronic acid, 98%, Thermo Scientific Chemicals
Catalog Number 15472049
Click to view available options
Quantity:
250 mg
1 g
5 g
351422-72-5
C11H10BNO2
199.02
MFCD16658918
SSVQXHWHJJERAP-UHFFFAOYSA-N
3-pyridin-3-yl phenyl boronic acid, 3-3-pyridyl benzeneboronic acid, 3-pyridin-3-yl phenylboronic acid, 3-3-pyridinyl phenylboronic acid, 3-3-pyridinyl phenyl boronic acid, boronic acid, 3-3-pyridinyl phenyl, 3-3-pyridyl phenylboronic acid, 3-pyridin-3-yl phenyl boronic acid pinacol ester
58526507
OB(O)C1=CC(=CC=C1)C1=CN=CC=C1
351422-72-5
C11H10BNO2
199.02
MFCD16658918
SSVQXHWHJJERAP-UHFFFAOYSA-N
3-pyridin-3-yl phenyl boronic acid, 3-3-pyridyl benzeneboronic acid, 3-pyridin-3-yl phenylboronic acid, 3-3-pyridinyl phenylboronic acid, 3-3-pyridinyl phenyl boronic acid, boronic acid, 3-3-pyridinyl phenyl, 3-3-pyridyl phenylboronic acid, 3-pyridin-3-yl phenyl boronic acid pinacol ester
58526507
OB(O)C1=CC(=CC=C1)C1=CN=CC=C1
It is used as an OLED material. Phenylboronic acids are efficient phenylating agents in copper promoted CN and CO bond cross-coupling reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as an OLED material. Phenylboronic acids are efficient phenylating agents in copper promoted CN and CO bond cross-coupling reactions.
Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Color White
Quantity 250 mg
Beilstein 20520996
IUPAC Name [3-(pyridin-3-yl)phenyl]boronic acid
Formula Weight 199.02
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3-(3-Pyridyl)benzeneboronic acid