missing translation for 'onlineSavingsMsg'
Learn More

3,3-Diphenylpropylamine, 97%

Product Code. p-7026757
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11430028 25 g 25g
11440028 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11430028 Supplier Thermo Scientific Alfa Aesar Supplier No. B22953.14
Add to basket
This item is not returnable. View return policy

CAS: 5586-73-2 | C15H17N | 211.308 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5586-73-2
Molecular Formula C15H17N
Molecular Weight (g/mol) 211.308
MDL Number MFCD00008202
InChI Key KISZTEOELCMZPY-UHFFFAOYSA-N
Synonym 3,3-diphenylpropylamine, benzenepropanamine, .gamma.-phenyl, 3-3-diphenylpropylamine, 3,3-diphenylpropan-1-amin, acmc-209lpx, 3.3-diphenylpropylamine, 3,3 diphenylpropylamine, 3,3-diphenyl propylamine, 3,3-diphenyl-propylamine, 3,3-diphenylpropylamine e
PubChem CID 79698
IUPAC Name 3,3-diphenylpropan-1-amine
SMILES C1=CC=C(C=C1)C(CCN)C2=CC=CC=C2

Specifications

Melting Point 24°C to 29°C
Boiling Point 166°C (2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.583
Quantity 25 g
Beilstein 1958595
Formula Weight 211.31
Percent Purity 97%
Chemical Name or Material 3,3-Diphenylpropylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.