missing translation for 'onlineSavingsMsg'
Learn More

3,3-Diphenyl-L-alanine, 95%, Thermo Scientific Chemicals

Product Code. p-7034483
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15478707

Brand: Thermo Scientific Alfa Aesar H52169.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

3,3-Diphenyl-L-alanine is used as organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3-Diphenyl-L-alanine is used as organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149597-92-2
Molecular Formula C15H15NO2
Molecular Weight (g/mol) 241.29
MDL Number MFCD01631989
InChI Key PECGVEGMRUZOML-AWEZNQCLSA-N
Synonym 3,3-diphenyl-l-alanine, s-2-amino-3,3-diphenylpropanoic acid, l-3,3-diphenylalanine, 3,3-diphenylalanine, 2s-2-amino-3,3-diphenylpropanoic acid, s-2-amino-3,3-diphenyl-propionic acid, 2s-2-amino-3,3-diphenyl-propanoic acid, d-3,3'-biphenylalanine, beta-phenylphenylalanine, pubchem18701
PubChem CID 162977
IUPAC Name (2S)-2-amino-3,3-diphenylpropanoic acid
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)C(C(=O)O)N

Specifications

UN Number UN2811
Quantity 250 mg
Formula Weight 241.29
Percent Purity 95%
Chemical Name or Material 3,3-Diphenyl-L-alanine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.