Learn More
DWK Life Sciences Kimble Chase™ 3.3 Borosilicate Glass Test Tubes
Product Code. 11823380
Change view
Click to view available options
Diameter (Metric):
16 mm
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
11823380 16 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11823380 Supplier DWK Life Sciences Supplier No. A613016001221
  • ONLINE EXCLUSIVE PRICE
    335.00 NOK
    301.59 NOK / Pack of 100
    Save 33.41 NOK
    9% Off
Estimated Shipment: 17-12-2026
Use round-bottom 3.3 Borosilicate Glass Test Tubes for laboratory needs when heat resistance is critical. These disposable test tubes are thick-walled and autoclavable.
  • Disposable
  • Borosilicate glass
  • Excellent resistance to temperature changes
  • Autoclavable
Specifications
Design Type Round bottom
Length (Metric) 130 mm
Packaging 100Pack
Diameter (Metric) 16 mm
Product Type Test Tube