Learn More
DWK Life Sciences Kimble Chase™ 3.3 Borosilicate Glass Test Tubes
Product Code. 11317544
Change view
Click to view available options
Length (Metric):
100 mm
130 mm
150 mm
180 mm
200 mm
Diameter (Metric):
10 mm
14 mm
16 mm
18 mm
20 mm
25 mm
30 mm
Packaging:
100Pack
1Each
50Pack
Unit Size:
Pack of 100
Pack of 50
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Packaging unitSize
11317544 150 mm 20 mm 100Pack Pack of 100
11863380 100 mm 10 mm 1Each Pack of 100
11862373 130 mm 14 mm 100Pack Pack of 100
11882373 130 mm 16 mm 100Pack Pack of 100
11337544 180 mm 18 mm 100Pack Pack of 100
11802383 180 mm 20 mm 100Pack Pack of 100
11812363 150 mm 25 mm 50Pack Pack of 50
11822363 200 mm 25 mm 50Pack Pack of 50
10676423 200 mm 30 mm 50Pack Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 11317544 Supplier DWK Life Sciences Supplier No. A615020001211
Only null left
Add to basket
Add to basket
Use round-bottom 3.3 Borosilicate Glass Test Tubes for laboratory needs when heat resistance is critical. These disposable test tubes are thick-walled and autoclavable.
  • Disposable
  • Borosilicate glass
  • Excellent resistance to temperature changes
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Design Type Round bottom
Length (Metric) 150 mm
Packaging 100Pack
Diameter (Metric) 20 mm
Product Type Test Tube