Learn More
DWK Life Sciences Kimble Chase™ 3.3 Borosilicate Glass Test Tubes
missing translation for 'orderingAttributeHoverText'
Diameter (Metric):
10 mm
missing translation for 'unitSize'
Pack of 100
Description
- Disposable
- Borosilicate glass
- Excellent resistance to temperature changes
- Autoclavable
Specifications
Specifications
| Design Type | Round bottom |
| Length (Metric) | 100 mm |
| Packaging | 100Pack |
| Diameter (Metric) | 10 mm |
| Product Type | Test Tube |