Learn More
Lenz Laborglasinstrumente™ 3.3 Borosilicate Glass 2-Way Stopcock
Click to view available options
Plug Bore:
4 mm
Unit Size:
Each
Specifications
Specifications
| Type | 2-way Stopcock |
| Plug Bore | 4 mm |
| Material | Borosilicate Glass |
| Certifications/Compliance | DIN 12540-1 |
| Includes | PTFE plug and screw thread retaining device |