missing translation for 'onlineSavingsMsg'
Learn More

3-(3,5-Dichlorophenyl)propionic acid, 97%, Thermo Scientific™

Product Code. 15490448
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15490448 25 g 25g
15480448 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15490448 Supplier Thermo Scientific Alfa Aesar Supplier No. H54009.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95333-95-2
Molecular Formula C9H8Cl2O2
Molecular Weight (g/mol) 219.06
MDL Number MFCD04037332
InChI Key GQICJNXGCGRFBA-UHFFFAOYSA-N
Synonym 3-3,5-dichlorophenyl propanoic acid, benzenepropanoic acid, 3,5-dichloro, 3-3,5-dichloro-phenyl-propionic acid, 3-3,5-dichlorophenyl propionic acid, 3-3',5'-dichlorophenyl propanoic acid, acmc-20anl7, 3,5-dichloro-benzenepropanoic acid, 3-3,5-dichlorophenyl propanoicacid, 3-3,5-di-chlorophenyl propionic acid, 3-3,5-dichloro-phenyl-propionicacid
PubChem CID 819305
IUPAC Name 3-(3,5-dichlorophenyl)propanoic acid
SMILES OC(=O)CCC1=CC(Cl)=CC(Cl)=C1

Specifications

Melting Point 56°C to 60°C
Quantity 25 g
Solubility Information Slightly soluble in water.
Formula Weight 219.07
Percent Purity 97%
Chemical Name or Material 3-(3,5-Dichlorophenyl)propionic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.