missing translation for 'onlineSavingsMsg'
Learn More

3,3',4',7-Tetrahydroxyflavone, 96%, Thermo Scientific Chemicals

Catalog Number 11389045
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
528-48-3
C15H10O6
286.239
MFCD00006829
XHEFDIBZLJXQHF-UHFFFAOYSA-N
fisetin, viset, 5-desoxyquercetin, fustel, 2-3,4-dihydroxyphenyl-3,7-dihydroxy-4h-chromen-4-one, fisetholz, superfustel, cotinin, fietin, fustet
5281614
CHEBI:42567
2-(3,4-dihydroxyphenyl)-3,7-dihydroxychromen-4-one
C1=CC(=C(C=C1C2=C(C(=O)C3=C(O2)C=C(C=C3)O)O)O)O
This item is not returnable. View return policy
528-48-3
C15H10O6
286.239
MFCD00006829
XHEFDIBZLJXQHF-UHFFFAOYSA-N
fisetin, viset, 5-desoxyquercetin, fustel, 2-3,4-dihydroxyphenyl-3,7-dihydroxy-4h-chromen-4-one, fisetholz, superfustel, cotinin, fietin, fustet
5281614
CHEBI:42567
2-(3,4-dihydroxyphenyl)-3,7-dihydroxychromen-4-one
C1=CC(=C(C=C1C2=C(C(=O)C3=C(O2)C=C(C=C3)O)O)O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 330°C (decomposition)
Quantity 1 g
Beilstein 292829
Merck Index 14,4088
Formula Weight 286.24
Percent Purity 97%
Chemical Name or Material 3,3',4',7-Tetrahydroxyflavone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'