Learn More
3,3,3-Trifluoroalanine, TRC
Specifications
Specifications
| Chemical Name or Material | 3,3,3-Trifluoroalanine |
| CAS | 17463-43-3 |
| Molecular Formula | C3H4F3NO2 |
| Quantity | 500 mg |
| Synonym | DL-3,3,3-Trifluoro-alanine, 3,3,3-Trifluoro-DL-alanine, 2-Amino-3,3,3-trifluoropropanoic Acid, 3,3,3-Trifluoro-DL-alanine, 3,3,3-Trifluoroalanine, DL-3,3,3-Trifluoroalanine, DL-Trifluoroalanine, DL-alpha-Trifluoromethylglycine, beta,beta,beta-Trifluoro-DL-alanine |
| InChI Formula | InChI=1S/C3H4F3NO2/c4-3(5,6)1(7)2(8)9/h1H,7H2,(H,8,9) |
| SMILES | NC(C(=O)O)C(F)(F)F |
| IUPAC Name | 2-amino-3,3,3-trifluoropropanoic acid |
| Molecular Weight (g/mol) | 143.06 |
| Formula Weight | 143.0194 |