Learn More
Alfa Aesar™ 3-(2-Thienyl)acrylic acid, predominantly trans, 98+%
Product Code. 11424133
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Product Code. Quantity unitSize
11424133 50 g 50g
11414133 10 g 10g
2 options
Product Code. 11424133
Brand: Alfa Aesar™ A10342.18
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Chemical Identifiers
CAS 15690-25-2
Molecular Formula C7H6O2S
Molecular Weight (g/mol) 154.183
MDL Number MFCD00005455
InChI Key KKMZQOIASVGJQE-ONEGZZNKSA-N
Synonym 3-2-thienyl acrylic acid, 3-thiophen-2-yl acrylic acid, 2-thiopheneacrylic acid, 2e-3-thiophen-2-yl prop-2-enoic acid, 2e-3-2-thienyl acrylic acid, 2-thienylacrylic acid, e-3-thiophen-2-yl acrylic acid, 2-thiopheneacrylic acid, e, unii-ino5aty68d, 2-propenoic acid, 3-2-thienyl
PubChem CID 735981
IUPAC Name (E)-3-thiophen-2-ylprop-2-enoic acid
SMILES C1=CSC(=C1)C=CC(=O)O
Specifications
Melting Point 145°C to 148°C
Quantity 50 g
Beilstein 112053
Formula Weight 154.19
Percent Purity ≥98%
Chemical Name or Material 3- (2-Thienyl)acrylic acid, predominantly trans