Learn More
3-(2-Pyridyl)-L-alanine dihydrochloride, 95%, Thermo Scientific™
Product Code. 15450639
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. Quantity unitSize
15450639 250 mg 250mg
15430639 1 g 1g
15440639 5 g 5g
3 options
Product Code. 15450639
Brand: Thermo Scientific Alfa Aesar H63770.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1082692-96-3
MDL Number MFCD06809729
Synonym l-2-pyridylalanine, 3-2-pyridyl-l-alanine, s-2-amino-3-pyridin-2-yl propanoic acid, h-2-pal-oh, l-3-2-pyridyl-alanine, 3-2-pyridyl-alanine, 2'-pyridyl-l-ala, 2s-2-amino-3-pyridin-2-yl propanoic acid, 2-aza-d-phenylalanine, 3-pyridin-2-yl-d-alanine
PubChem CID 2761499
Specifications
Molecular Formula C8H10N2O2
InChI Key PDRJLZDUOULRHE-ZETCQYMHSA-N
SMILES C1=CC=NC(=C1)CC(C(=O)O)N
IUPAC Name (2S)-2-amino-3-pyridin-2-ylpropanoic acid
Molecular Weight (g/mol) 166.18
Quantity 250 mg
Formula Weight 239.1
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-(2-Pyridyl)-L-alanine dihydrochloride