Learn More
3-(2-Bromoethyl)-7-hydroxy-4-methyl-coumarin, TRC
Specifications
Specifications
| Chemical Name or Material | 3-(2-Bromoethyl)-7-hydroxy-4-methyl-coumarin |
| CAS | 10185-03-2 |
| Molecular Formula | C12H11BrO3 |
| Quantity | 100 mg |
| Synonym | 3-(2-Bromoethyl)-4-methyl-7-hydroxycoumarin, 3-(2-Bromoethyl)-7-hydroxy-4-methylcoumarin, 3-(2-Bromoethyl)-7-hydroxy-4-methyl-2H-1-benzopyran-2-one |
| InChI Formula | InChI=1S/C12H11BrO3/c1-7-9-3-2-8(14)6-11(9)16-12(15)10(7)4-5-13/h2-3,6,14H,4-5H2,1H3 |
| SMILES | CC1=C(CCBr)C(=O)Oc2cc(O)ccc12 |
| IUPAC Name | 3-(2-bromoethyl)-7-hydroxy-4-methylchromen-2-one |
| Molecular Weight (g/mol) | 283.12 |
| Formula Weight | 281.9892 |