Learn More
3-(2-Aminoethyl)-5-methylindole hydrochloride, 97%, Thermo Scientific Chemicals
Product Code. 15435728
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
15435728 5 g 5g
15425016 25 g 25g
2 options
Product Code. 15435728
Brand: Thermo Scientific Alfa Aesar H59415.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1010-95-3
Molecular Formula C11H15ClN2
Molecular Weight (g/mol) 210.71
MDL Number MFCD00012683
InChI Key RBHDFGBPJGEYCK-UHFFFAOYSA-N
Synonym 5-methyltryptamine hydrochloride, 2-5-methyl-1h-indol-3-yl ethanamine hydrochloride, 3-2-aminoethyl-5-methylindole hydrochloride, 5-methyl-1h-indole-3-ethylamine monohydrochloride, 5-methyltryptamine hcl, 1h-indole-3-ethanamine, 5-methyl-, monohydrochloride, 2-5-methylindol-3-yl ethylamine, chloride, 5-methyltryptaminehydrochloride, 5-methyl-1h-indole-3-ethylamine hcl, pubchem24287
PubChem CID 6917145
IUPAC Name hydrogen 2-(5-methyl-1H-indol-3-yl)ethan-1-amine chloride
SMILES [H+].[Cl-].CC1=CC=C2NC=C(CCN)C2=C1
Specifications
Melting Point 290°C to 292°C (decomposition)
Quantity 5 g
Beilstein 3707780
Sensitivity Hygroscopic
Formula Weight 210.71
Percent Purity 97%
Chemical Name or Material 3-(2-Aminoethyl)-5-methylindole hydrochloride