Learn More
3-(2,2,2-Trifluoroethoxy)prop-1-yne, TRC
Specifications
Specifications
| Chemical Name or Material | 3-(2,2,2-Trifluoroethoxy)prop-1-yne |
| CAS | 1481-06-7 |
| Molecular Formula | C5H5F3O |
| Quantity | 25 mg |
| Synonym | 3-(2,2,2-Trifluoroethoxy)-1-propyne, 2-Propynyl 2,2,2-trifluoroethyl Ether |
| InChI Formula | InChI=1S/C5H5F3O/c1-2-3-9-4-5(6,7)8/h1H,3-4H2 |
| SMILES | FC(F)(F)COCC#C |
| IUPAC Name | 3-(2,2,2-Trifluoroethoxy)-1-propyne |
| Molecular Weight (g/mol) | 138.09 |
| Formula Weight | 138.03 |