missing translation for 'onlineSavingsMsg'
Learn More

3-(1h-1,2,4-Triazol-1-ylmethyl)benzaldehyde, 97%, Thermo Scientific™

Product Code. p-4510671
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. Quantity unitSize
10307483 250 mg 250mg
10719215 1 g 1g
10123134 5 g 5g
3 options
This item is not returnable. View return policy

Product Code. 10307483

Brand: Thermo Scientific Maybridge CC45404CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 876316-30-2
Molecular Formula C10H9N3O
Molecular Weight (g/mol) 187.202
InChI Key IBVURYHFXBPNCW-UHFFFAOYSA-N
Synonym 3-1h-1,2,4-triazol-1-ylmethyl benzaldehyde, benzaldehyde,3-1h-1,2,4-triazol-1-ylmethyl, 3-1,2,4-triazol-1-ylmethyl benzaldehyde, 3-1h-1,2,4-triazol-1-yl methyl benzaldehyde, 3-1,2,4-triazolylmethyl benzaldehyde
PubChem CID 22360585
IUPAC Name 3-(1,2,4-triazol-1-ylmethyl)benzaldehyde
SMILES C1=CC(=CC(=C1)CN2C=NC=N2)C=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 187.2
Percent Purity 97%
Chemical Name or Material 3-(1H-1, 2, 4-Triazol-1-ylmethyl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.