Learn More
2xYT Agar (Powder/Molecular Genetics), Fisher BioReagents
Specifications
Specifications
| Additive | 2xyt Agar |
| Form | Powder |
| For Use With (Application) | Molecular Genetics |
| Packaging Type | Poly Bottle |
| Quantity | 2000 g |