missing translation for 'onlineSavingsMsg'
Learn More

(2S,3S)-(+)-Di-O-benzoyltartaric Acid, Sigma-Aldrich™

Product Code. 15800661
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15800661 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15800661 Supplier Sigma Aldrich Supplier No. 8188690025
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For resolution of racemates for synthesis

TRUSTED_SUSTAINABILITY

Specifications

CAS 17026-42-5
Melting Point 150°C to 154°C
Quantity 25 g
Molecular Formula C6H5COOCH(COOH)CH(COOH)OCOC6H5
MDL Number MFCD00063222
Synonym (2S,3S)-(+)-Di-O-benzoyltartaric acid, Di-O-benzoyl-D-tartaric acid, (2S,3S)-(+)-Di-O-benzoyltartaric acid
Molecular Weight (g/mol) 358.3
Chemical Name or Material (2S,3S)-(+)-Di-O-Benzoyltartaric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.