Learn More
(2S,3aS,7aS)-1-[(2S)-2-[[(1S)-1-Carboxybutyl]amino]propanoyl]octahydro-1H-indole-2-carboxylic Acid (Perindoprilat), Mikromol
Description
(2S,3aS,7aS)-1-[(2S)-2-[[(1S)-1-Carboxybutyl]amino]propanoyl]octahydro-1H-indole-2-carboxylic Acid (Perindoprilat), Mikromol
Specifications
Specifications
| Chemical Name or Material | Perindoprilat |
| CAS | 95153-31-4 |
| Analyte or Component Names | Perindoprilat |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H28 N2 O5 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1H-Indole-2-carboxylic acid, 1-[(2S)-2-[[(1S)-1-carboxybutyl]amino]-1-oxopropyl]octahydro-, (2S,3aS,7aS)-, 1H-Indole-2-carboxylic acid, 1-[2-[(1-carboxybutyl)amino]-1-oxopropyl]octahydro-, [2S-[1[R*(R*)],2α,3aβ,7aβ]]-, Perindoprilat, S 9780, Perindopril RC B; |
| InChI Formula | InChI=1S/C17H28N2O5/c1-3-6-12(16(21)22)18-10(2)15(20)19-13-8-5-4-7-11(13)9-14(19)17(23)24/h10-14,18H,3-9H2,1-2H3,(H,21,22)(H,23,24)/t10-,11-,12-,13-,14-/m0/s1 |
| Solution Type | Neat |
| SMILES | CCC[C@H](N[C@@H](C)C(=O)N1[C@H]2CCCC[C@H]2C[C@H]1C(=O)O)C(=O)O |
| Show More |