Learn More
(2S,2'S)-2,2'-([1,1'-Biphenyl]-4,4'-diyldi-1H-imidazole-5,2-diyl)bis-1-pyrrolidinecarboxylic Acid 1,1'-Bis(1,1-dimethylethyl) Ester, TRC
Product Code. 30332750
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30332750 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30332750 Supplier Toronto Research Chemicals Supplier No. TRCB39815525MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (2S,2'S)-2,2'-([1,1'-Biphenyl]-4,4'-diyldi-1H-imidazole-5,2-diyl)bis-1-pyrrolidinecarboxylic Acid 1,1'-Bis(1,1-dimethylethyl) Ester
CAS 1007882-23-6
Molecular Formula C36 H44 N6 O4
Quantity 25 mg
InChI Formula InChI=1S/C36H44N6O4/c1-35(2,3)45-33(43)41-19-7-9-29(41)31-37-21-27(39-31)25-15-11-23(12-16-25)24-13-17-26(18-14-24)28-22-38-32(40-28)30-10-8-20-42(30)34(44)46-36(4,5)6/h11-18,21-22,29-30H,7-10,19-20H2,1-6H3,(H,37,39)(H,38,40)/t29-,30-/m0/s1
SMILES CC(C)(C)OC(=O)N1CCC[C@H]1c2ncc([nH]2)c3ccc(cc3)c4ccc(cc4)c5cnc([nH]5)[C@@H]6CCCN6C(=O)OC(C)(C)C
IUPAC Name tert-butyl (2S)-2-[5-[4-[4-[2-[(2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidin-2-yl]-1H-imidazol-5-yl]phenyl]phenyl]-1H-imidazol-2-yl]pyrrolidine-1-carboxylate
Molecular Weight (g/mol) 624.772
Formula Weight 624.342