Learn More
(2S)-2-Amino-5-(carbamoylamino)pentanoic Acid (Citrulline), Mikromol™
Description
(2S)-2-Amino-5-(carbamoylamino)pentanoic Acid (Citrulline)
Specifications
Specifications
| Chemical Name or Material | L-Citrulline |
| CAS | 372-75-8 |
| Analyte or Component Names | L-Citrulline |
| Shipping Condition | Room Temperature |
| Molecular Formula | C6 H13 N3 O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | N5-(Aminocarbonyl)-L-ornithine, Ornithine, N5-carbamoyl-, L- (8CI), Cit, Citrulline, L-(+)-Citrulline, L-Citrulline, N5-Carbamoyl-L-ornithine, NSC 27425, Ndelta-Carbamylornithine, alpha-Amino-delta-ureidovaleric acid, delta-Ureidonorvaline, Arginine Impurity B |
| InChI Formula | InChI=1S/C6H13N3O3/c7-4(5(10)11)2-1-3-9-6(8)12/h4H,1-3,7H2,(H,10,11)(H3,8,9,12)/t4-/m0/s1 |
| SMILES | N[C@@H](CCCNC(=O)N)C(=O)O |
| IUPAC Name | (2S)-2-amino-5-(carbamoylamino)pentanoic acid |
| Show More |