Learn More
(2RS)-2-(2-Fluorobiphenyl-4-yl)-2-hydroxypropanoic Acid, Mikromol™
Product Code. 30050618
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Product Code. 30050618
Brand: Mikromol MM0727.03
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2RS)-2-(2-Fluorobiphenyl-4-yl)-2-hydroxypropanoic Acid
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (2RS)-2-(2-Fluorobiphenyl-4-yl)-2-hydroxypropanoic Acid
CAS 61466-95-3
Analyte or Component Names (2RS)-2-(2-Fluorobiphenyl-4-yl)-2-hydroxypropanoic Acid
Shipping Condition Room Temperature
Molecular Formula C15 H13 F O3
Certifications/Compliance ISO 9001
Synonym (2RS)-2-(2-Fluorobiphenyl-4-yl)-2-hydroxypropanoic Acid, Flurbiprofen Imp. C (EP), Flurbiprofen Impurity C
InChI Formula InChI=1S/C15H13FO3/c1-15(19,14(17)18)11-7-8-12(13(16)9-11)10-5-3-2-4-6-10/h2-9,19H,1H3,(H,17,18)
SMILES CC(O)(C(=O)O)c1ccc(c(F)c1)c2ccccc2
IUPAC Name 2-(3-fluoro-4-phenylphenyl)-2-hydroxypropanoic acid
Molecular Weight (g/mol) 260.26
Quantity 50 mg
Formula Weight 260.0849 g/mol