Learn More
(2R)-N-(2-Benzoyl-4-chlorophenyl)-1-(phenylmethyl)-2-pyrrolidinecarboxamide Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | (2R)-N-(2-Benzoyl-4-chlorophenyl)-1-(phenylmethyl)-2-pyrrolidinecarboxamide Hydrochloride |
| CAS | 1021462-91-8 |
| Molecular Formula | C25H23ClN2O2 . (HCl) |
| Quantity | 100 mg |
| Synonym | (2R)-N-(2-Benzoyl-4-chlorophenyl)-1-(phenylmethyl)-2-pyrrolidinecarboxamide Hydrochloride |
| InChI Formula | InChI=1S/C25H23ClN2O2.ClH/c26-20-13-14-22(21(16-20)24(29)19-10-5-2-6-11-19)27-25(30)23-12-7-15-28(23)17-18-8-3-1-4-9-18;/h1-6,8-11,13-14,16,23H,7,12,15,17H2,(H,27,30);1H/t23-;/m1./s1 |
| SMILES | O=C([C@H]1CCCN1CC2=CC=CC=C2)NC3=C(C=C(C=C3)Cl)C(C4=CC=CC=C4)=O.Cl |
| IUPAC Name | (2R)-N-(2-benzoyl-4-chlorophenyl)-1-benzylpyrrolidine-2-carboxamide;hydrochloride |
| Molecular Weight (g/mol) | 455.38 |
| Formula Weight | 454.12 |