missing translation for 'onlineSavingsMsg'
Learn More

(2R,3R)-(+)-Tartaric Acid, Sigma-Aldrich™

Product Code. 15860461
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15860461 2.5 kg 2.5kg
15840461 100 g 100g
15850461 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15860461 Supplier Sigma Aldrich Supplier No. 8185312500
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For resolution of racemates for synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-69-4
Molecular Formula HOOCCH(OH)CH(OH)COOH
Molecular Weight (g/mol) 150.09

Specifications

Melting Point 168°C to 170°C
Density 1.76 g/cm3 at 20°C
Quantity 2.5 kg
MDL Number MFCD00064207
Synonym (2R,3R)-(+)-Tartaric acid, 2,3-Dihydroxybutanedioic acid, (2R,3R)-(+)-Tartaric acid
Chemical Name or Material (2R,3R)-(+)-Tartaric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.