Learn More
(2R,3R)-(-)-Di-O-4-toluoyl-L-tartaric acid, Sigma-Aldrich™
Specifications
Specifications
| Chemical Name or Material | (2R,3R)-(-)-Di-O-4-toluoyl-L-tartaric acid |
| CAS | 32634-66-5 |
| Melting Point | 165°C to 171°C |
| Quantity | 100 g |
| Molecular Formula | (4-(CH3)C6H4COOCHCOOH)2 |
| Synonym | (2R,3R)-(-)-Di-O-4-toluoyl-L-tartaric acid, Di-O-4-toluoyl-L-tartaric acid, (2R,3R)-(-)-Di-O-4-toluoyl-L-tartaric acid |
| Molecular Weight (g/mol) | 386.35 |
| Grade | Not Available |