Learn More
(2R,3R)-2-Ethyl-3-(hydroxymethyl)-4-(1-methyl-1H-imidazol-5-yl)butanoic Acid Sodium Salt (Isopilocarpic Acid Sodium Salt), Mikromol™
Description
(2R,3R)-2-Ethyl-3-(hydroxymethyl)-4-(1-methyl-1H-imidazol-5-yl)butanoic Acid Sodium Salt (Isopilocarpic Acid Sodium Salt)
Specifications
Specifications
| Chemical Name or Material | (2R,3R)-2-Ethyl-3-(hydroxymethyl)-4-(1-methyl-1H-imidazol-5-yl)butanoic Acid Sodium Salt (Isopilocarpic Acid Sodium Salt) |
| CAS | 101769-87-3 |
| Analyte or Component Names | (2R,3R)-2-Ethyl-3-(hydroxymethyl)-4-(1-methyl-1H-imidazol-5-yl)butanoic Acid Sodium Salt (Isopilocarpic Acid Sodium Salt) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C11 H17 N2 O3 . Na |
| Certifications/Compliance | ISO 9001 |
| Synonym | Pilocarpine Hydrochloride Imp. C (EP), Pilocarpine Imp. C (EP), 1H-Imidazole-5-butanoic acid, alpha-ethyl-beta-(hydroxymethyl)-1-methyl-, monosodium salt, [R-(R*,R*)]- (9CI), (2R,3R)-2-Ethyl-3-(hydroxymethyl)-4-(1-methyl-1H-imidazol-5-yl)butanoic Acid Sodium Salt, Pilocarpine Hydrochloride Imp. C (EP) as Sodium Salt, Isopilocarpic Acid Sodium Salt, Pilocarpine Hydrochloride Impurity C as Sodium Salt, Pilocarpine Nitrate Impurity C as Sodium Salt, Pilocarpine Impurity C as Sodium Salt, Pilocarpine Impurity C as Sodium Salt |
| InChI Formula | InChI=1S/C11H18N2O3.Na/c1-3-10(11(15)16)8(6-14)4-9-5-12-7-13(9)2;/h5,7-8,10,14H,3-4,6H2,1-2H3,(H,15,16);/q;+1/p-1/t8-,10+;/m0./s1 |
| SMILES | [Na+].CC[C@H]([C@H](CO)Cc1cncn1C)C(=O)[O-] |
| IUPAC Name | sodium;(2R,3R)-2-ethyl-3-(hydroxymethyl)-4-(3-methylimidazol-4-yl)butanoate |
| Show More |