Learn More
(2R,3R)-2-Aminobutane-1,3-diol, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 3228-51-1 |
| Molecular Formula | C4H11NO2 |
| MDL Number | MFCD00191173 |
| Quantity | 10 g |
| SMILES | C[C@@H](O)[C@H](N)CO |
| Molecular Weight (g/mol) | 105.14 |
| Percent Purity | 95% |
| Chemical Name or Material | (2R,3R)-2-Aminobutane-1,3-diol |