Learn More
(2R)-2-Amino-3-(4-aminophenyl)propanoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | (2R)-2-Amino-3-(4-aminophenyl)propanoic Acid |
| CAS | 102281-45-8 |
| Molecular Formula | C9H12N2O2 |
| Quantity | 500 mg |
| Synonym | 4-Amino-D-phenylalanine, D-4-Aminophenylalanine, p-Amino-D-phenylalanine |
| InChI Formula | InChI=1S/C9H12N2O2/c10-7-3-1-6(2-4-7)5-8(11)9(12)13/h1-4,8H,5,10-11H2,(H,12,13)/t8-/m1/s1 |
| SMILES | N[C@H](Cc1ccc(N)cc1)C(=O)O |
| IUPAC Name | (2R)-2-amino-3-(4-aminophenyl)propanoic acid |
| Molecular Weight (g/mol) | 180.2 |
| Formula Weight | 180.0899 |