missing translation for 'onlineSavingsMsg'
Learn More

TM5441, MedChemExpress

Product Code. 16480056
Encompass
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16480056 2 mg 2mg
16486277 5 mg 5mg
16479966 50 mg 50mg
16454666 100 mg 100mg
16453558 10 mg 10mg
16422776 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16480056 Supplier Medchem Express Supplier No. HY101761

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress TM5441 is an orally bioavailable inhibitor of plasminogen activator inhibitor-1 (PAI-1), has IC50 values between 13.9 and 51.1 μM and induces intrinsic apoptosis in several human cancer cell lines. TM5441 attenuates Nω-nitro-l-arginine methyl ester-induced cardiac hypertension and vascular senescence.

TM5441 is an orally bioavailable inhibitor of plasminogen activator inhibitor-1 (PAI-1), has IC50 values between 13.9 and 51.1 μM and induces intrinsic apoptosis in several human cancer cell lines. TM5441 attenuates Nω-nitro-l-arginine methyl ester-induced cardiac hypertension and vascular senescence.

Specifications

Chemical Name or Material TM5441
CAS 1190221-43-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H17ClN2O6
Quantity 2 mg
Solubility Information DMSO : ≥ 300 mg/mL (699.59 mM)
SMILES ClC1=CC=C(NC(COCC(NC2=CC=CC(C3=COC=C3)=C2)=O)=O)C(C(O)=O)=C1
Molecular Weight (g/mol) 428.82
Formula Weight 428.82
Percent Purity 98.18%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.