Learn More
Thermo Scientific™ Barcode Reader for KingFisher™ Duo Prime
Product Code. 15347317 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15347317 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15347317 Supplier Thermo Scientific™ Supplier No. N16640
Estimated Shipment: 16-11-2026
Track samples and reagents quickly and easily with the Barcode Reader for Thermo Scientific KingFisher Duo Prime.
The Barcode Reader for Thermo Scientific KingFisher Duo Prime is an optional accessory that allows users to track samples directly in the instrument’s internal software quickly and easily.
The Barcode Reader for Thermo Scientific KingFisher Duo Prime is an optional accessory that allows users to track samples directly in the instrument’s internal software quickly and easily.
TRUSTED_SUSTAINABILITY
Specifications
Format 1D and 2D
Description 2D Barcode reader for KingFisher
For Use With (Equipment) KingFisher Duo Prime
Quantity Each