Learn More
3M™ 2840 Series Polycarbonate Lens Safety Glasses
Specifications
Specifications
| Certifications/Compliance | EN 166:2001 |
| Lens Coating | Anti-fog, Scratch-resistant |
| Lens Tint | Clear |
| Product Type | Safety glasses; Safety Spectacles |
| For Use With (Application) | Construction, pharmaceuticals |
| Frame Color | Silver |
| No. per Pack | 20/Pk. |
| Temple Style | Adjustable length |
| Lens Material | Polycarbonate |