Learn More
27-Hydroxycholesterol, MedChemExpress
Product Code. 16492589
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Product Code. Quantity unitSize
16492589 1 mg 1mg
16461718 5 mg 5mg
2 options
Product Code. 16492589
Brand: Medchem Express HYN2371
MedChemExpress 27-Hydroxycholesterol is a selective estrogen receptor modulator and an agonist of the liver X receptor.
27-Hydroxycholesterol is a selective estrogen receptor modulator and an agonist of the liver X receptor.
Specifications
Chemical Name or Material 27-Hydroxycholesterol
CAS 20380-11-4
Color White
Purity Grade Notes Research
Molecular Formula C27H46O2
Quantity 1 mg
Solubility Information Ethanol : 16.67 mg/mL (41.40 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (2.48 mM; Need ultrasonic)
SMILES C[C@@H](CO)CCC[C@@H](C)[C@H]1CC[C@@]2([H])[C@]3([H])CC=C4C[C@@H](O)CC[C@]4(C)[C@@]3([H])CC[C@]12C
Molecular Weight (g/mol) 402.65
Formula Weight 402.65
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death