missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human WISP-1/CCN4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB1627-SP
This item is not returnable.
View return policy
Description
WISP-1/CCN4 Monoclonal specifically detects WISP-1/CCN4 in Human samples. It is validated for Western Blot.
Specifications
| WISP-1/CCN4 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| O95388 | |
| WISP1 | |
| Mouse myeloma cell line NS0-derived recombinant human WISP-1/CCN4 Thr23-Asn367 Accession # O95388 | |
| 25 μg | |
| Primary | |
| Detects human WISP-1/CCN4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human (rh) CTGF, rhNOV, or rmWISP-1. | |
| Human | |
| Purified |
| Western Blot | |
| 213611 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CCN family member 4, CCN4, CCN4WISP1tc, WISP1, WISP-1, WISP1c, WISP1i, WNT1 induced secreted protein 1, Wnt-1 inducible signaling pathway protein 1, WNT1 inducible signaling pathway protein 1, wnt-1 signaling pathway protein 1, Wnt-1-induced secreted protein, WNT1-inducible-signaling pathway protein 1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8840 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction